3-Amino-2-pyridone 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Amino-2-hydroxypyridine, 3-Amino-2-pyridinol, 3-Amino-2-pyridinone
360710010 360710050
ACRO360710010EA 1425 SEK
ACRO360710010 ACRO360710050
3-Amino-2-pyridone 97%
3-Amino-2-pyridone
Formel: C₅H₆N₂O
MW: 110,12 g/mol
Smältpunkt: 118…130 °C
Storage Temperature: Omgivande
MDL Number: MFCD03411556
CAS nummer: 33630-99-8
Order Now
Specifikation testresultat
Appearance Dark brown to black Powder
Infrared spectrum Conforms
HPLC ≥96.0 %