3-Bromo-2,5-diaminopyridine 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Bromopyridine-2,5-diamine, 2,5-Diamino-3-bormopyridine, 3-Bromanylpyridine-2,5-diamine
360660010 360660050
ACRO360660010EA 1500 SEK
ACRO360660010 ACRO360660050
3-Bromo-2,5-diaminopyridine 99%
3-Bromo-2,5-diaminopyridine
Formel: C₅H₆BrN₃
MW: 188,028 g/mol
Smältpunkt: 117…122 °C
Storage Temperature: Omgivande
MDL Number: MFCD03427651
CAS nummer: 896160-69-3
Order Now
Specifikation testresultat
Appearance Dark brown to violet Powder
Infrared spectrum Conforms
HPLC ≥98.5 %