Bis(neopentyl glycolato)diboron 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 5,5,5',5'-Tetramethyl-2,2'-bi(1,3,2-dioxaborinane), 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane
358890010 358890050
ACRO358890010EA 995 SEK
ACRO358890010 ACRO358890050
Bis(neopentyl glycolato)diboron 98%
Bis(neopentyl glycolato)diboron
Formel: C₁₀H₂₀B₂O₄
Smältpunkt: 178…183 °C
Storage Temperature: Omgivande
MDL Number: MFCD02093062
CAS nummer: 201733-56-4
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Infrared spectrum Conforms
GC ≥97.5 %