Orotic acid, vattenfri 98%

Leverantör: Thermo Scientific
Varning

Synonyms: 2,6-Dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid

352880100 352881000
ACRO352880100EA 690 SEK
ACRO352880100 ACRO352881000
Orotic acid, vattenfri 98%
Orotic acid
Formel: C₅H₄N₂O₄
MW: 156,10 g/mol
Smältpunkt: >300 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00006027
CAS nummer: 65-86-1

Order Now

Specifikation testresultat

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤2 % (K.F.)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR