4-Chloro-3-sulphamoylbenzoic acid 98%
Leverantör: Thermo Fisher Scientific
Varning
303220010 303220050
ACRO303220010EA 745 SEK
ACRO303220010 ACRO303220050
4-Chloro-3-sulphamoylbenzoic acid 98%
4-Chloro-3-sulphamoylbenzoic acid
Formel: C₇H₆ClNO₄S
MW: 235.65 g/mol
Kokpunkt: 256…258 °C (760 mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD00012375
CAS nummer: 1205-30-7
Order Now
Specifikation testresultat
Appearance White Powder
Infrared spectrum Conforms
Melting point 258 to 263 °C
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure