Kaliumhydroxid 1 N i vatten
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Potash caustic, Caustic potash
278400010 278400025 278400250
ACRO278400010EA 615 SEK
ACRO278400010 ACRO278400025 ACRO278400250
Kaliumhydroxid 1 N i vatten
Kaliumhydroxid
Formel: KOH
MW: 56.11 g/mol
Kokpunkt: 100 °C (760 mmHg)
Smältpunkt: 360
Densitet: 1.05 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00003553
CAS nummer: 1310-58-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806
Order Now
Specifikation testresultat
Appearance Clear colorless Solution
Strength 1 N Exact strength on the certificate of analysis