Kaliumklorid 99.999%
Leverantör: Thermo Fisher Scientific
Synonyms: Potassiumchloride
268960250 268961000
ACRO268960250EA 1975 SEK
ACRO268960250 ACRO268961000
Kaliumklorid 99.999%
Kaliumklorid
Formel: KCl
MW: 74.55 g/mol
Kokpunkt: 1420 °C (760 mmHg)
Smältpunkt: 770 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011360
CAS nummer: 7447-40-7
Merck Index: 14,07621
Order Now
Specifikation testresultat
Appearance White Crystalline powder or crystals
Total trace metal impurities ≤10 ppm