cis-1,2-Diaminocyclohexane 97%
Leverantör: Thermo Scientific
Fara
Synonyms: cis-Cyclohexane-1,2-diamine, (±)-cis-1,2-Diaminocyclohexane
248660010 248660050
ACRO248660010EA 1375 SEK
ACRO248660010 ACRO248660050
cis-1,2-Diaminocyclohexane 97%
cis-1,2-Diaminocyclohexane
Formel: C₆H₁₄N₂
MW: 114,19 g/mol
Kokpunkt: 92 °C (14mbar)
Densitet: 0,952 g/cm³
Flampunkt: 70 °C
Storage Temperature: Omgivande
MDL Number: MFCD00063746
CAS nummer: 1436-59-5
UN: 2735
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Colorless to brown Liquid (May darken on storage)
Infrared spectrum Conforms
GC ≥96.0 %