Propyl chloroformate 98%
Leverantör: Thermo Fisher Scientific
Fara
245291000 245290250
ACRO245291000EA 3425 SEK
ACRO245291000 ACRO245290250
Propyl chloroformate 98%
Propyl chloroformate
Formel: C₄H₇ClO₂
Smältpunkt: –70 °C
Densitet: 1,09 g/cm³
Flampunkt: 16 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000649
CAS nummer: 109-61-5
UN: 2740
ADR: 6.1,I
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4030 to 1.4050 (20°C, 589 nm)
GC ≥97.5 %