3-Methyl-3-oxetanemethanol 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Methyl-3-oxethanemethanol
240840100 240840500
ACRO240840100EA 675 SEK
ACRO240840100 ACRO240840500
3-Methyl-3-oxetanemethanol 98%
3-Methyl-3-oxetanemethanol
Formel: C₅H₁₀O₂
MW: 102,13 g/mol
Kokpunkt: 122 °C (70 mmHg)
Densitet: 1,034 g/cm³
Flampunkt: 97 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00010273
CAS nummer: 3143-02-0
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %