L-Cysteic acid 99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: (R)-2-Amino-3-sulphopropanoic acid
226120100 226120500
ACRO226120100EA 1350 SEK
ACRO226120100 ACRO226120500
L-Cysteic acid 99%
L-Cysteic acid
Formel: C₃H₇NO₅S
MW: 169.15 g/mol
Smältpunkt: 267…269 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007524
CAS nummer: 498-40-8
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Solubility (5 % in water) Clear colorless
Specific optical rotation +7° to +9° (20°C, 589 nm) (c=5, H2O)
Titration with NaOH ≥98.5 %