2-Methylbenzofuran 96%
Leverantör: Thermo Scientific
Varning
208400010 208400050
ACRO208400010EA 1725 SEK
ACRO208400010 ACRO208400050
2-Methylbenzofuran 96%
2-Methylbenzofuran
Formel: C₉H₈O
MW: 132,16 g/mol
Kokpunkt: 197…198 °C (760 mmHg)
Densitet: 1,057 g/cm³
Flampunkt: 67 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005850
CAS nummer: 4265-25-2
Order Now
Specifikation testresultat
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥95.0 %