α-Chlorothioanisole 97%
Leverantör: Thermo Scientific
Synonyms: Chloromethyl phenyl sulphide
202360050 202360250
ACRO202360050EA 1300 SEK
ACRO202360050 ACRO202360250
α-Chlorothioanisole 97%
α-Chlorothioanisole
Formel: C₇H₇ClS
Kokpunkt: 66 °C (,3hPa)
Densitet: 1,23 g/cm³
Flampunkt: 101 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000921
CAS nummer: 7205-91-6
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %