Ammoniumsulfat 99.9995%
Leverantör: Thermo Fisher Scientific
Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
194150500 194152500
ACRO194150500EA 1625 SEK
ACRO194150500 ACRO194152500
Ammoniumsulfat 99.9995%
Ammonium sulphate
Formel: (NH₄)₂SO₄
MW: 132.14 g/mol
Smältpunkt: 280 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003391
CAS nummer: 7783-20-2
Merck Index: 13,00559
Order Now
Specifikation testresultat
Appearance White Crystalline powder or crystals and/or chunks
Total trace metal impurities ≤5 ppm
Gravimetry 23.7 to 24.9 % (S)