Kobolt 99.995%, powder and/or granules
Leverantör: Thermo Fisher Scientific
Fara
192850050 192850250
ACRO192850050EA 1500 SEK
ACRO192850050 ACRO192850250
Kobolt 99.995%, powder and/or granules
Kobolt
Formel: Co
MW: 58.93 g/mol
Kokpunkt: 2870 °C (760 mmHg)
Smältpunkt: 1495 °C
Storage Temperature: Omgivande
MDL Number: MFCD00010935
CAS nummer: 7440-48-4
Merck Index: 12,02488
Order Now
Specifikation testresultat
Appearance Grey Powder and/or granules
Total trace metal impurities ≤50 ppm