2-(2-Hydroxyphenyl)ethanol 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-(2-Hydroxyethyl)phenol, 2-Hydroxyphenethyl alcohol
182490010 182490050
ACRO182490010EA 680 SEK
ACRO182490010 ACRO182490050
2-(2-Hydroxyphenyl)ethanol 98%
2-(2-Hydroxyphenyl)ethanol
Formel: C₈H₁₀O₂
MW: 138,17 g/mol
Kokpunkt: 168…169 °C (2 mmHg)
Densitet: 1,159 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00002890
CAS nummer: 7768-28-7
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Clear yellow to yellow-brown Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5560 to 1.5600 (20°C, 589 nm)
GC ≥97.5 %
HPLC ≥96.0 %
Specific optical rotation -125° to -142° (20°C, 589 nm) (0.25 % in Methanol)