N-Methyl-N-prop-2-ynylamine 98+%

Leverantör: Thermo Fisher Scientific
Fara

165540010 165540050
ACRO165540010EA 840 SEK
ACRO165540010 ACRO165540050
N-Methyl-N-prop-2-ynylamine 98+%
N-Methyl-N-prop-2-ynylamine
Formel: C₄H₇N
Kokpunkt: 82…84 °C (760 mmHg)
Densitet: 0,83 g/cm³
Flampunkt: 1 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00008573
CAS nummer: 35161-71-8
UN: 2733
ADR: 3,II

Order Now

Specifikation testresultat

Appearance Clear colorless to yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4370 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR