2-Amino-4-methoxybenzothiazole 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4-Methoxybenzothiazol-2-ylamine
160350010 160350050 160350250
ACRO160350010EA 735 SEK
ACRO160350010 ACRO160350050 ACRO160350250
2-Amino-4-methoxybenzothiazole 97%
2-Amino-4-methoxybenzothiazole
Formel: C₈H₈N₂OS
MW: 180.23 g/mol
Kokpunkt: 335
Smältpunkt: 153…155 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005792
CAS nummer: 5464-79-9
Order Now
Specifikation testresultat
Appearance Beige Crystalline powder
Infrared spectrum Conforms
Melting point 150 to 153 °C
GC ≥96.0 %