1,2,3,6-Tetrahydropyridine 98%
Leverantör: Thermo Scientific
Fara
158870010 158870050
ACRO158870010EA 2200 SEK
ACRO158870010 ACRO158870050
1,2,3,6-Tetrahydropyridine 98%
1,2,3,6-Tetrahydropyridine
Formel: C₅H₉N
MW: 83,13 g/mol
Kokpunkt: 108 °C (760 mmHg)
Smältpunkt: –48 °C
Densitet: 0,91 g/cm³
Flampunkt: 16 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005949
CAS nummer: 694-05-3
UN: 2410
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow to tan Liquid
Infrared spectrum Conforms
Refractive index 1.4790 to 1.4830 (20°C, 589 nm)
GC ≥97.5 %
Sulfate (SO4) ≤0.01 %
Iron (Fe) ≤0.002 %
Sodium (Na) ≤0.005 %
Insoluble matter ≤0.01 % (in water)