5-Quinolinol 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 5-Hydroxyquinoline, Quinolin-5-ol
156430010 156430050
ACRO156430010EA 1375 SEK
ACRO156430010 ACRO156430050
5-Quinolinol 97%
5-Quinolinol
Formel: C₉H₇NO
MW: 145,16 g/mol
Kokpunkt: 313 °C (1013 hPa)
Smältpunkt: 223…226 °C
Densitet: 1,57 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00006792
CAS nummer: 578-67-6
Order Now
Specifikation testresultat
Appearance White to yellow to beige to brown Powder or crystals
Infrared spectrum Conforms
HPLC ≥96.0 %