N,N'-Diethylethylenediamine 95%
Leverantör: Thermo Fisher Scientific
Fara
155520050 155520250
ACRO155520050EA 640 SEK
ACRO155520050 ACRO155520250
N,N'-Diethylethylenediamine 95%
N,N'-Diethylethylenediamine
Formel: C₆H₁₆N₂
MW: 116.21 g/mol
Kokpunkt: 152…154 °C (760 mmHg)
Smältpunkt: –11 °C
Densitet: 0.811 g/cm³
Flampunkt: 33 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009033
CAS nummer: 111-74-0
UN: 2685
ADR: 8 II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4315 to 1.4335 (20°C, 589 nm)
GC ≥94 %
NMR (1H-NMR) Consistent with structure