Cyclohexanecarboxaldehyde 97%

Leverantör: Thermo Fisher Scientific
Varning

148670250 148671000
ACRO148670250EA 1075 SEK
ACRO148670250 ACRO148671000
Cyclohexanecarboxaldehyde 97%
Cyclohexanecarboxaldehyde
Formel: C₇H₁₂O
Kokpunkt: 161…163 °C (760 mmHg)
Densitet: 0,926 g/cm³
Flampunkt: 41 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001457
CAS nummer: 2043-61-0
UN: 1989
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Clear colorless to light pink Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4520 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR