ω-Bromo-2-methoxyacetophenone 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 2-Methoxyphenacyl bromide, α-Bromo-2-methoxyacetophenone, 2-Bromo-2'-methoxyacetophenone, 2-Bromo-1-(2-methoxyphenyl)ethanone, 2-Bromo-1-(2-methoxyphenyl)ethan-1-one, Bromomethyl o-methoxyphenyl ketone
146270010 146270050 146270250
ACRO146270010EA 605 SEK
ACRO146270010 ACRO146270050 ACRO146270250
ω-Bromo-2-methoxyacetophenone 98%
ω-Bromo-2-methoxyacetophenone
Formel: C₉H₉BrO₂
Kokpunkt: 130 °C (1 mmHg)
Smältpunkt: 43…45 °C
Flampunkt: 113 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000196
CAS nummer: 31949-21-0
UN: 3261
ADR: 8,II
Order Now
Specifikation testresultat
Appearance White to brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 42 to 47 °C
GC ≥97.5 %