3-Aminoisoxazole 95%
Leverantör: Thermo Scientific
Varning
133530050 133530250
ACRO133530050EA 1900 SEK
ACRO133530050 ACRO133530250
3-Aminoisoxazole 95%
3-Aminoisoxazole
Formel: C₃H₄N₂O
MW: 84,08 g/mol
Kokpunkt: 226…228 °C (760 mmHg)
Densitet: 1,38 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00038814
CAS nummer: 1750-42-1
Order Now
Specifikation testresultat
Appearance Clear yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5090 to 1.5120 (20°C, 589 nm)
GC ≥94 %