2-(Methylthio)aniline 97%
Leverantör: Thermo Fisher Scientific
Fara
127110050 127110250
ACRO127110050EA 635 SEK
ACRO127110050 ACRO127110250
2-(Methylthio)aniline 97%
2-(Methylthio)aniline
Formel: C₇H₉NS
Kokpunkt: 234 °C (760 mmHg)
Densitet: 1,111 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007708
CAS nummer: 2987-53-3
UN: 2810
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Light yellow to brown Liquid
Infrared spectrum Conforms
GC ≥98.5 %
Water ≤0.06 %