Methyl-5-chloro-5-oxovalerate 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Methyl glutaryl chloride, Methyl 4-chloroformylbutyrate
126540100 126540500
ACRO126540100EA 930 SEK
ACRO126540100 ACRO126540500
Methyl-5-chloro-5-oxovalerate 98%
Methyl-5-chloro-5-oxovalerate
Formel: C₆H₉ClO₃
MW: 164,59 g/mol
Kokpunkt: 110 °C (17 mmHg)
Densitet: 1,191 g/cm³
Flampunkt: 82 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000756
CAS nummer: 1501-26-4
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4470 (20°C, 589 nm)
GC ≥97.5 %