Methoxyacetonitrile 98%
Leverantör: Thermo Fisher Scientific
Varning
125660050 125660250 125661000
ACRO125660050EA 1175 SEK
ACRO125660050 ACRO125660250 ACRO125661000
Methoxyacetonitrile 98%
Methoxyacetonitrile
Formel: C₃H₅NO
Kokpunkt: 118…119 °C (731 mmHg)
Densitet: 0,95 g/cm³
Flampunkt: 31 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001892
CAS nummer: 1738-36-9
UN: 1993
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3800 to 1.3830 (20°C, 589 nm)
GC ≥97.5 %