Bis(2-mercaptoethyl)sulphide 90%
Leverantör: Thermo Fisher Scientific
Synonyms: 2,2'-Thiodiethanethiol, 2-Mercaptoethyl Sulphide
125490100 125490500
ACRO125490100EA 1225 SEK
ACRO125490100 ACRO125490500
Bis(2-mercaptoethyl)sulphide 90%
Bis(2-mercaptoethyl)sulphide
Formel: C₄H₁₀S₃
MW: 154.30 g/mol
Kokpunkt: 135…136 °C (10 mmHg)
Smältpunkt: –11 °C
Densitet: 1.183 g/cm³
Flampunkt: 90 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004893
CAS nummer: 3570-55-6
UN: 3334
ADR: 9 III
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5900 to 1.5980 (20°C, 589 nm)
GC ≥88 %
Non-volatile matter ≤0.001 %