1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,2',2'',2'''-(((Ethane-1,2-diylbis(oxy))bis(2,1-phenylene))bis(azanetriyl))tetraacetic acid, BAPTA
123310010 123310050 123311000
ACRO123310010EA 2425 SEK
ACRO123310010 ACRO123310050 ACRO123311000
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid 97%
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid
Formel: C₂₂H₂₄N₂O₁₀
MW: 476,44 g/mol
Smältpunkt: 177…179 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00036255
CAS nummer: 85233-19-8
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %