2-Cyclohexen-1-ol 95%
Leverantör: Thermo Fisher Scientific
Varning
111230010 111230050 111230100
ACRO111230010EA 1025 SEK
ACRO111230010 ACRO111230050 ACRO111230100
2-Cyclohexen-1-ol 95%
2-Cyclohexen-1-ol
Formel: C₆H₁₀O
Kokpunkt: 164…166 °C (760 mmHg)
Densitet: 0,984 g/cm³
Flampunkt: 58 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001570
CAS nummer: 822-67-3
UN: 1987
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥94 %
Titration with HClO4 ≥97.5 %