Calcium pantothenate, pulver NF, USP
Leverantör: Spectrum Chemical
Synonyms: Calcium D-Pantothenate, D-Pantothenic acid calcium salt, D-Calcium pantothenate, Calcium (R)-3-(2,4-dihydroxy-3,3-dimethylbutanamido)propanoate, Vitamin B5 calcium salt, Calcium D-(+)-pantothenate
CA159-2.5KG CA159-50KGBL
SPCMCA159-2.5KGEA 34375 SEK
SPCMCA159-2.5KG SPCMCA159-50KGBL
Calcium pantothenate, pulver NF, USP
Calcium pantothenate
Formel: C₁₈H₃₂CaN₂O₁₀
MW: 476.54 g/mol
MDL Number: MFCD00002766
CAS nummer: 137-08-6
EINECS: 205-278-9