Triisopropyl borate 98+%, AcroSeal®
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Tri-i-propylborate
427141000 427148000
427141000.EA 1275 SEK
427141000. 427148000.
Triisopropyl borate 98+%, AcroSeal®
Triisopropyl borate
Formel: C₉H₂₁BO₃
MW: 188.07 g/mol
Kokpunkt: 139
Smältpunkt: –59 °C
Densitet: 0.814 g/cm³
Flampunkt: 17 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008872
CAS nummer: 5419-55-6
UN: 2616
ADR: 3 II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3755 to 1.3775 (20°C, 589 nm)
GC ≥98.0 %