(4-Bromophenoxy)-tert-butyldimethylsilane 97%
Leverantör: Thermo Scientific
Fara
Synonyms: 1-Bromo-4-(tert-butyldimethylsiloxy)benzene, (4-Bromophenoxy)(tert-butyl)dimethylsilane
427090050 427090250
ACRO427090050EA 1175 SEK
ACRO427090050 ACRO427090250
(4-Bromophenoxy)-tert-butyldimethylsilane 97%
(4-Bromophenoxy)-tert-butyldimethylsilane
Appearance: Clear colorless to light yellow Liquid
Formel: C₁₂H₁₉BrOSi
MW: 287,272 g/mol
Kokpunkt: 137 °C (25 mmHg)
Densitet: 1,174 g/cm³
Flampunkt: 12 °C
Storage Temperature: Omgivande
MDL Number: MFCD00239382
CAS nummer: 67963-68-2
UN: 1993
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure