1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene 90%
Leverantör: Thermo Scientific
Fara
Synonyms: tert-Butyl 4-(aminomethyl)benzylcarbamate
426900010 426900050
ACRO426900010EA 1225 SEK
ACRO426900010 ACRO426900050
1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene 90%
1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene
Appearance: White to brown Crystalline powder or solid
Formel: C₁₃H₂₀N₂O₂
MW: 236,3100 g/mol
Smältpunkt: 64…68 °C
Storage Temperature: Omgivande
MDL Number: MFCD02683058
CAS nummer: 108468-00-4
UN: 3263
ADR: 8,III
Order Now
Specifikation testresultat
Appearance White to brown Crystalline powder or solid
Infrared spectrum Conforms
GC ≥89.0 %
NMR (1H-NMR) Consistent with structure