2-(Diethylamino)ethyl bromide hydrobromide 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-Bromotriethylamine Hydrobromide, 2-Bromoethyldiethylamine Hydrobromide, 2-Bromo-N,N-diethylethylamine Hydrobromide
426860250 426861000
ACRO426860250EA 2125 SEK
ACRO426860250 ACRO426861000
2-(Diethylamino)ethyl bromide hydrobromide 98%
2-(Diethylamino)ethyl bromide hydrobromide
Appearance: White to yellow Powder or solid
Formel: C₆H₁₄BrN
MW: 261,00 g/mol
Smältpunkt: 211…215 °C
Storage Temperature: Omgivande
MDL Number: MFCD00040376
CAS nummer: 1069-72-3
UN: 2811
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance White to yellow Powder or solid
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
NMR (1H-NMR) Consistent with structure