Kalium-tert-butanolat 20% (w/w) 1.6-1.7 M i THF, AcroSeal®, ren
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Potassium tert-butylate, Potassium tert-butanolate
428791000 428798000
428791000.EA 3850 SEK
428791000. 428798000.
Kalium-tert-butanolat 20% (w/w) 1.6-1.7 M i THF, AcroSeal®, ren
Kalium-tert-butanolat
Formel: C₄H₉KO
MW: 112.21 g/mol
Flampunkt: –21 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012162
CAS nummer: 865-47-4
UN: 2920
ADR: 8 I
Order Now
Specifikation testresultat
Appearance Light yellow to amber Solution
Strength 1.60 to 1.75 M