N-(tert-Butoxycarbonyl)-1,4-diaminobutane 95%
Leverantör: Thermo Scientific
Fara
Synonyms: N-(4-Aminobutyl)carbamic Acid tert-Butyl Ester, N-(tert-Butoxycarbonyl)-1,4-butanediamine, NH2-C4-NH-Boc, N-Boc-1,4-diaminobutane, N-Boc-1,4-butanediamine, tert-Butyl N-(4-Aminobutyl)carbamate, tert-Butyl (4-aminobutyl)carbamate
426190010 426190050
ACRO426190010EA 1300 SEK
ACRO426190010 ACRO426190050
N-(tert-Butoxycarbonyl)-1,4-diaminobutane 95%
N-(tert-Butoxycarbonyl)-1,4-diaminobutane
Appearance: Clear colorless to light yellow Liquid
Formel: C₉H₂₀N₂O₂
MW: 188,27 g/mol
Densitet: 0,984 g/cm³
Flampunkt: 109 °C
Storage Temperature: Omgivande
MDL Number: MFCD00210019
CAS nummer: 68076-36-8
UN: 2735
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94 %
NMR consistent with structure