DSS (Disuccinimidyl suberate) 97%
Leverantör: Thermo Scientific
Varning
Synonyms: DSS
426020010 426020050
ACRO426020010EA 2550 SEK
ACRO426020010 ACRO426020050
DSS (Disuccinimidyl suberate) 97%
DSS (Disuccinimidyl suberate)
Formel: C₁₆H₂₀N₂O₈
MW: 368,34 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00049059
CAS nummer: 68528-80-3
Order Now
Specifikation testresultat
Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure