Sulphur trioxide N,N-dimethylformamide complex 47+% SO3_aktiv
Leverantör: Thermo Fisher Scientific
Fara
425850050 425850250
ACRO425850050EA 805 SEK
ACRO425850050 ACRO425850250
Sulphur trioxide N,N-dimethylformamide complex 47+% SO3_aktiv
Sulphur trioxide N,N-dimethylformamide complex
Formel: C₃H₇NO₄S
MW: 153.152 g/mol
Smältpunkt: 155…158 °C
Flampunkt: >120 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00043411
CAS nummer: 29584-42-7
EINECS: 249-701-5
UN: 3261
ADR: 8 II
Order Now
Specifikation testresultat
Appearance Beige to light brown Crystals
Infrared spectrum Conforms
Titration with NaOH ≥47.0 % (as SO3)