DL-3-Pyrrolidinol 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Pyrrolidin-3-ol, 3-Pyrrolidinol, DL-3-Hydroxypyrrolidine
425750010 425750050
ACRO425750010EA 1900 SEK
ACRO425750010 ACRO425750050
DL-3-Pyrrolidinol 97%
DL-3-Pyrrolidinol
Appearance: Clear colorless to yellow Liquid
Formel: C₄H₉NO
Kokpunkt: 179 °C (760 mmHg)
Densitet: 1,076 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005256
CAS nummer: 40499-83-0
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
GC ≥96.0 %