n-Octylmagnesium bromide 2 M i Dietyleter, AcroSeal®
Leverantör: Thermo Fisher Scientific
Fara
427811000 427818000
427811000.EA 1350 SEK
427811000. 427818000.
n-Octylmagnesium bromide 2 M i Dietyleter, AcroSeal®
n-Octylmagnesium bromide
Appearance: Yellow to brown Solution
Formel: C₈H₁₇BrMg
Kokpunkt: 35 °C (760 mmHg)
Densitet: 0,961 g/cm³
Flampunkt: –35 °C
Storage Temperature: Omgivande
MDL Number: MFCD00191566
CAS nummer: 17049-49-9
UN: 2924
ADR: 3,I
Order Now
Specifikation testresultat
Appearance Yellow to brown Solution
Titration Grignard 1.9 to 2.2 M
GC ≥98.0 %