(±)-2-Metyltetrahydrofuran, extra dry over Molecular Sieve 99+% stabilised, AcroSeal®
Leverantör: Thermo Fisher Scientific
Fara
396630010 396631000
396630010.EA 4150 SEK
396630010. 396631000.
(±)-2-Metyltetrahydrofuran, extra dry over Molecular Sieve 99+% stabilised, AcroSeal®
(±)-2-Metyltetrahydrofuran
Formel: C₅H₁₀O
Kokpunkt: 79 °C (1013 hPa)
Smältpunkt: –136 °C
Densitet: 0,86 g/cm³
Flampunkt: –11 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00005367
CAS nummer: 96-47-9
UN: 2536
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.002 %
GC ≥99.0 %
Stabilizer 150 to 400 ppm BHT
Peroxides ≤50 ppm