Bis(tricyclohexylphosphine)palladium(II) dichloride 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: trans-Dichlorobis(tricyclohexylphosphine)palladium(II), Dichlorobis(tricyclohexylphosphine)palladium(II)
395890010 395892500
ACRO395890010EA 1950 SEK
ACRO395890010 ACRO395892500
Bis(tricyclohexylphosphine)palladium(II) dichloride 98%
Bis(tricyclohexylphosphine)palladium(II) dichloride
Appearance: Yellow Powder
Formel: C₃₆H₆₆Cl₂P₂Pd
MW: 738.19 g/mol
Smältpunkt: 270 °C
Storage Temperature: Omgivande
MDL Number: MFCD00191830
CAS nummer: 29934-17-6
Order Now
Specifikation testresultat
Appearance Yellow Powder
Infrared spectrum Conforms
Elemental Analysis ≥97.5 %