Allylboronic acid pinacol ester 98%
Leverantör: Thermo Fisher Scientific
Varning
395130010 395130050
ACRO395130010EA 795 SEK
ACRO395130010 ACRO395130050
Allylboronic acid pinacol ester 98%
Allylboronic acid pinacol ester
Formel: C₉H₁₇BO₂
Kokpunkt: 50…53 °C (5 mmHg)
Flampunkt: 46 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00013347
CAS nummer: 72824-04-5
UN: 1993
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥97.5 %