(2S,3S)-N-tert-Boc-3-amino-1,2-epoxy-4-phenylbutane 98%
Leverantör: Thermo Scientific
Varning
Synonyms: (2S,3S)-N-t-Boc-3-amino-1,2-epoxy-4-phenylbutane, (2S,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, Darunavir Impurity 22
395020010 395020050 395020250
ACRO395020010EA 2950 SEK
ACRO395020010 ACRO395020050 ACRO395020250
(2S,3S)-N-tert-Boc-3-amino-1,2-epoxy-4-phenylbutane 98%
(2S,3S)-N-tert-Boc-3-amino-1,2-epoxy-4-phenylbutane
Appearance: White to off-white Powder
Formel: C₁₅H₂₁NO₃
MW: 263,34 g/mol
Smältpunkt: 125…127 °C
Storage Temperature: Kylskåp
MDL Number: MFCD02258997
CAS nummer: 98737-29-2
UN: 3077
ADR: 9,III
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %