(4-(Benzyloxy)phenyl)hydrazine hydrochloride 98%
Leverantör: Thermo Scientific
Varning
394030010 394030050
ACRO394030010EA 995 SEK
ACRO394030010 ACRO394030050
(4-(Benzyloxy)phenyl)hydrazine hydrochloride 98%
(4-(Benzyloxy)phenyl)hydrazine hydrochloride
Appearance: Cream to brown Powder
Formel: C₁₃H₁₅ClN₂O
MW: 250,7240 g/mol
Smältpunkt: 187…189 °C
Storage Temperature: Omgivande
MDL Number: MFCD00137741
CAS nummer: 52068-30-1
UN: 2811
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Cream to brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %