Diphosphoryl tetrachloride 90+%
Leverantör: Thermo Scientific
Fara
Synonyms: Diphosphoryl chloride, Pyrophosphoryl chloride
393620050 393620250
ACRO393620050EA 500 SEK
ACRO393620050 ACRO393620250
Diphosphoryl tetrachloride 90+%
Diphosphoryl tetrachloride
Formel: Cl₄O₃P₂
MW: 251,76 g/mol
Kokpunkt: 90 °C (12 mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD00061542
CAS nummer: 13498-14-1
UN: 3264
ADR: 8,I
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Titration with NaOH ≥90.0 %