4-Bromo-1-butanol 85+%
Leverantör: Thermo Fisher Scientific
Fara
390010010 390010050 390010250
ACRO390010010EA 915 SEK
ACRO390010010 ACRO390010050 ACRO390010250
4-Bromo-1-butanol 85+%
4-Bromo-1-butanol
Formel: BrH₂C(CH₂)₂CH₂OH
Kokpunkt: 56…58 °C (2 mmHg)
Flampunkt: 7 °C
Storage Temperature: Frys
MDL Number: MFCD00015387
CAS nummer: 33036-62-3
UN: 1987
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless to purple to brown Liquid
Infrared spectrum Conforms
GC ≥85.0 %