Kvicksilver(I)sulfat 97%
Leverantör: Thermo Scientific
Fara
Synonyms: Mercurous sulphate, Dimercury sulphate
389920050 389921000
ACRO389920050EA 500 SEK
ACRO389920050 ACRO389921000
Kvicksilver(I)sulfat 97%
Kvicksilver(I)sulfat
Formel: Hg₂O₄S
MW: 497,24 g/mol
Kokpunkt: 356,6 °C (1013 hPa)
Smältpunkt: –38,9 °C
Densitet: 7,56 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00016138
CAS nummer: 7783-36-0
UN: 1645
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance White to yellow to beige Powder
Titration Iodimetric 96.0 to 104.0 %