(S)-(-)-2,3-Epoxy-1-propanol 97%, 98% ee
Leverantör: Thermo Scientific
Fara
Synonyms: (S)-(-)-Glycidol, (S)-Oxiran-2-ylmethanol
411000010 411000050 411000250
ACRO411000010EA 655 SEK
ACRO411000010 ACRO411000050 ACRO411000250
(S)-(-)-2,3-Epoxy-1-propanol 97%, 98% ee
(S)-(-)-2,3-Epoxy-1-propanol
Formel: C₃H₆O₂
MW: 74,08 g/mol
Smältpunkt: –54 °C
Flampunkt: 66 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00074874
CAS nummer: 60456-23-7
UN: 2810
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4340 (20°C, 589 nm)
GC ≥96.0 %
Enantiomeric excess ≥97.5 % (GC)
Specific optical rotation -12° to -18° (20°C, 589 nm) (neat)